Learn More
Waters Corp Waters Polypropylene Vial, 15 x 45 mm, 3 mL Volume
Catalog No. 50780525 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This polypropylene-molded vial is an economical, non-reactive alternative to glass, ideal for samples that may adhere to polar surfaces. Its design allows for cremation while closed, enhancing safety during disposal.
  • Clean, polypropylene-molded vial.
  • Prevents compound adherence common with glass surfaces.
  • Cost-effective alternative to glass.
  • Non-reactive plastic.
  • Permits cremation while closed, reducing hazardous exposure.
  • Features a universal screw cap.
  • Forms an excellent, mechanically crushed seal when capped.
  • Septum is mechanically fixed during piercing.
  • No special tools required for assembly.
  • Available with Pre-slit PTFE/Silicone or Polyethylene Septumless Cap options.
  • Inspected and tested for quality and cleanliness.
  • Compatible with all autosamplers.
  • Chemically resistant.
Catalog No. 50-780-525 Supplier Waters Corp Supplier No. 186000999
This polypropylene-molded vial is an economical, non-reactive alternative to glass, ideal for samples that may adhere to polar surfaces. Its design allows for cremation while closed, enhancing safety during disposal.
  • Clean, polypropylene-molded vial.
  • Prevents compound adherence common with glass surfaces.
  • Cost-effective alternative to glass.
  • Non-reactive plastic.
  • Permits cremation while closed, reducing hazardous exposure.
  • Features a universal screw cap.
  • Forms an excellent, mechanically crushed seal when capped.
  • Septum is mechanically fixed during piercing.
  • No special tools required for assembly.
  • Available with Pre-slit PTFE/Silicone or Polyethylene Septumless Cap options.
  • Inspected and tested for quality and cleanliness.
  • Compatible with all autosamplers.
  • Chemically resistant.