Learn More
S2 Media 15x60mm Malt Extract Agar (MEA) - Contact Plates, 10/pack
Catalog No. 502995859
Encompass_Preferred
Malt Extract Agar (MEA) is a selective and non-differential medium used for the isolation, cultivation, and enumeration of yeasts and molds. MEA was first described by Thom and Church in 1926 in a study of Aspergillus spp. claiming the high carbohydrate content ensured rapid growth. • Product meets BAM and USP-NF reference methods and performance criteria.• MEA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.
This item is not returnable. View return policy
Catalog No. 50-299-5859 Supplier S2 Media Supplier No. 5331
Malt Extract Agar (MEA) is a selective and non-differential medium used for the isolation, cultivation, and enumeration of yeasts and molds. MEA was first described by Thom and Church in 1926 in a study of Aspergillus spp. claiming the high carbohydrate content ensured rapid growth. • Product meets BAM and USP-NF reference methods and performance criteria.• MEA should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.