Learn More
S2 Media 15x60mm Tryptic Soy Agar with Lecithin and Tween 80 (TSA w/LT), 10/pack
Catalog No. 502995927
Encompass_Preferred
Tryptic Soy Agar with Lecithin and Tween80 (TSA w/LT) is a non-selective and non-differential medium utilized for the detection and enumeration of microorganisms present on surface of sanitary importance. Prepared plates are utilized for environmental monitoring within biologically controlled environments. These plates are suitable for active and passive air sampling as well as for personnel monitoring of finger tips (Finger Dabs). Lecithin is utilized to neutralize quaternary ammonium compounds. Polysorbate (Tween) 80 is used to neutralize substituted phenolic disinfectants. • Product meets USP-NF reference methods and performance criteria.• This product utilizes a friction lock lid. • TSB w/ LT should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Microbial Content Test Agar.
Catalog No. 50-299-5927 Supplier S2 Media Supplier No. 5144
Tryptic Soy Agar with Lecithin and Tween80 (TSA w/LT) is a non-selective and non-differential medium utilized for the detection and enumeration of microorganisms present on surface of sanitary importance. Prepared plates are utilized for environmental monitoring within biologically controlled environments. These plates are suitable for active and passive air sampling as well as for personnel monitoring of finger tips (Finger Dabs). Lecithin is utilized to neutralize quaternary ammonium compounds. Polysorbate (Tween) 80 is used to neutralize substituted phenolic disinfectants. • Product meets USP-NF reference methods and performance criteria.• This product utilizes a friction lock lid. • TSB w/ LT should be used by trained professionals within a laboratory setting. • Product is not intended for use in the diagnosis or treatment of disease or other human conditions.• Also known as Microbial Content Test Agar.