Learn More
Medchemexpress LLC 16:0-18:0-16:0 TG-d5 | 944709-25-5 | 99.9% | 840.40 | 1 MG
SDP
Catalog No. 5000363649
Encompass_Preferred
16:0-18:0-16:0 TG-d5 is a deuterium labeled 16:0-18:0-16:0 TG. It is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs. The product is a solid, white to off-white in color.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-63649 Supplier Medchemexpress LLC Supplier No. HY146914S1MG
16:0-18:0-16:0 TG-d5 is a deuterium labeled 16:0-18:0-16:0 TG. It is intended for research use only. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, primarily as tracers for quantitation during the drug development process. Deuteration can potentially affect the pharmacokinetic and metabolic profiles of drugs. The product is a solid, white to off-white in color.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS