Learn More
Avanti Polar Lipids 16:0-18:1 PA, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphate (sodium salt), 169437-35-8, MFCD00672609, 25mg
Catalog No. NC1123091
Encompass
Empirical Formula (Hill Notation): C37H70O8PNa, Molecular Weight: 696.91, Purity >99%, Storage Temperature: -20°C.
Catalog No. NC1123091 Supplier Avanti Polar Lipids Supplier No. 840857C25MG
Empirical Formula (Hill Notation): C37H70O8PNa, Molecular Weight: 696.91, Purity >99%, Storage Temperature: -20°C.