Learn More
Medchemexpress LLC 16:0-18:1 PA-d31 sodium | 474943-59-4 | 99.0% | 728.10 | 1 MG
SDP
Catalog No. 5000363912
Encompass_Preferred
16:0-18:1 PA-d31 (sodium) is deuterium labeled 16:0-18:1 PA (sodium). It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-003-63912 Supplier Medchemexpress LLC Supplier No. HY146896S1MG
16:0-18:1 PA-d31 (sodium) is deuterium labeled 16:0-18:1 PA (sodium). It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Deuteration has potential to affect the pharmacokinetic and metabolic profiles of drugs.