Learn More
Avanti Polar Lipids 1-palmitoyl-2-oleoyl-glycero-3-phosphocholine, 26853-31-6, MFCD00077387, 100 mg
Catalog No. NC1102319
Encompass
Molecular Formula: C42H82NO8P, Formula Weight: 760.076, Purity >99%. POPC is typically considered one of the model lipids for biophysical experiments. The fatty acid composition, i.e., saturated chain in the sn-1 position and unsaturated chain in the sn-2 position, mimics mammalian phospholipid composition. Since the major constituent in Egg PC is 1-palmitoyl-2-oleoyl PC, POPC is an excellent synthetic substitute for EPC.
Catalog No. NC1102319 Supplier Avanti Polar Lipids Supplier No. 850457C1G
Molecular Formula: C42H82NO8P, Formula Weight: 760.076, Purity >99%. POPC is typically considered one of the model lipids for biophysical experiments. The fatty acid composition, i.e., saturated chain in the sn-1 position and unsaturated chain in the sn-2 position, mimics mammalian phospholipid composition. Since the major constituent in Egg PC is 1-palmitoyl-2-oleoyl PC, POPC is an excellent synthetic substitute for EPC.