Learn More
Avanti Polar Lipids 16:0-18:1 PE, 26662-94-2, 25mg
Catalog No. NC9641068
Encompass
Molecular Formula C39H76NO8P, Purity >99%, Formula Weight 717.996, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine
Catalog No. NC9641068 Supplier Avanti Polar Lipids Supplier No. 850757C25MG
Molecular Formula C39H76NO8P, Purity >99%, Formula Weight 717.996, 1-palmitoyl-2-oleoyl-sn-glycero-3-phosphoethanolamine