Learn More
Medchemexpress LLC 16:0-18:1 PE-d31 | 326495-44-7 | 99.8% | 749.19 | 1 MG
SDP
Catalog No. 5000361494
Encompass_Preferred
16:0-18:1 PE-d31 is deuterium labeled 16:0-18:1 PE. It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-003-61494 Supplier Medchemexpress LLC Supplier No. HY146882S1MG
16:0-18:1 PE-d31 is deuterium labeled 16:0-18:1 PE. It is for research use only and not sold to patients. Stable heavy isotopes of hydrogen, carbon, and other elements have been incorporated into drug molecules, largely as tracers for quantitation during the drug development process. Deuteration has gained attention because of its potential to affect the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.