Learn More
Medchemexpress LLC 16:0-18:1 PG-d5 | 1246298-34-9 | 98.0% | 771.07 | 1 MG
SDP
Catalog No. 5000360675
Encompass_Preferred
16:0-18:1 PG-d5 is deuterium labeled 16:0-18:1 PG. It is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-003-60675 Supplier Medchemexpress LLC Supplier No. HY146878S1MG
16:0-18:1 PG-d5 is deuterium labeled 16:0-18:1 PG. It is for research use only and not sold to patients. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS