Learn More
Medchemexpress LLC 16:0-18:1 PS-d31 sodium | 327178-96-1 | 99.08% | 815.18 | 1 MG
SDP
Supplier:  Medchemexpress LLC HY146885S1MG
Encompass_Preferred
16:0-18:1 PS-d31 (sodium) is a deuterium labeled version of 16:0-18:1 PS (sodium). Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules for use as tracers in quantitation during drug development processes. Deuteration can impact the pharmacokinetic and metabolic profiles of drugs.
  • Deuterium labeled compound
  • Suitable as a tracer for quantitation in drug development
  • Potential to affect pharmacokinetic and metabolic profiles of drugs
Catalog No. 50-003-61495