Learn More
Medchemexpress LLC 16:0-18:1 PS-d31 sodium | 327178-96-1 | MFCD02259271 | 99.1% | 815.18 g/mol | C40H44NO10PNaD31 | 5 MG
SDP
Catalog No. 5000223103
Encompass_Preferred
16:0-18:1 PS-d31 sodium is a deuterium-labeled phosphatidylserine (PS) lipid supplied as the sodium salt for research use. The d31 isotopic label enables sensitive mass spectrometry tracing and quantification in lipidomics and metabolic studies. Supplied in small research quantities, it is intended for laboratory applications only.
  • Deuterium-labeled phosphatidylserine sodium salt for isotope tracing.
  • High purity (99.08%) suitable for analytical applications.
  • Optimized for mass spectrometry and lipidomics workflows.
  • Useful as an internal standard or tracer in metabolic studies.
  • Available in small research quantities for laboratory experiments.
Catalog No. 50-002-23103 Supplier Medchemexpress LLC Supplier No. HY146885S5MG
16:0-18:1 PS-d31 sodium is a deuterium-labeled phosphatidylserine (PS) lipid supplied as the sodium salt for research use. The d31 isotopic label enables sensitive mass spectrometry tracing and quantification in lipidomics and metabolic studies. Supplied in small research quantities, it is intended for laboratory applications only.
  • Deuterium-labeled phosphatidylserine sodium salt for isotope tracing.
  • High purity (99.08%) suitable for analytical applications.
  • Optimized for mass spectrometry and lipidomics workflows.
  • Useful as an internal standard or tracer in metabolic studies.
  • Available in small research quantities for laboratory experiments.