Learn More
Avanti Polar Lipids Hexadecyl Coenzyme A (ammonium salt), 71425-89-3, 5 mg
Catalog No. NC1243321
Encompass
Molecular Formula: C37H77N10O16P3S, Formula Weight: 1042.445, Purity >99%.
Catalog No. NC1243321 Supplier Avanti Polar Lipids Supplier No. 870740P5MG
Molecular Formula: C37H77N10O16P3S, Formula Weight: 1042.445, Purity >99%.