Learn More
Avanti Polar Lipids 1,2-dipalmitoyl-sn-glycero-3-phosphoethanolamine-N-(6-azidohexanoyl) (ammonium salt), 2252461-34-8, 25 mg
Catalog No. NC1008046
Encompass
Formula Weight: 848.145, Purity >99%, Storage Temperature: -20°C. This reagent is a modified lipid containing an omega-terminal azide. The terminal azide group can be used in a highly specific linking reaction with alkyne-containing reagents, known as ‘click chemistry’, in the presence of a copper (Cu)-containing catalyst.
Catalog No. NC1008046 Supplier Avanti Polar Lipids Supplier No. 870126P25MG
Formula Weight: 848.145, Purity >99%, Storage Temperature: -20°C. This reagent is a modified lipid containing an omega-terminal azide. The terminal azide group can be used in a highly specific linking reaction with alkyne-containing reagents, known as ‘click chemistry’, in the presence of a copper (Cu)-containing catalyst.