Learn More
Avanti Polar Lipids 1-palmitoyl-2-hydroxy-sn-glycero-3-phosphate (sodium salt), 17618-08-5, MFCD00674362, 25mg
Catalog No. NC1123093
Encompass
Molecular Formula: C19H38O7PNa, Formula Weight: 432.464, Purity>99%, Storage Temperature: -20°C.
Catalog No. NC1123093 Supplier Avanti Polar Lipids Supplier No. 857123P25MG
Molecular Formula: C19H38O7PNa, Formula Weight: 432.464, Purity>99%, Storage Temperature: -20°C.