Learn More
Avanti Polar Lipids 16:0 Lyso PC | 17364-16-8 | MFCD00036904 | 200mg
Supplier:  Avanti Polar Lipids 855675C200MG
Encompass
16:0 Lyso PC | Putity: >99% LPC; may contain up to 10% of the 2-LPC isomer | MW: 495.63 | 17364-16-8 | MFCD00036904 | 200mg
Catalog No. NC1368335