Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 16:0 monomethyl PE (1,2-NMeDPPE) | 3930-13-0 | 98.0% | 706.0 g/mol | C38H76NO8P | 10 MG
SDP

Catalog No. 5000284672
Encompass_Preferred

16:0 Monomethyl PE (1,2-NMeDPPE) is a monomethylated phosphatidylethanolamine lipid supplied as a purified powder for laboratory research applications. It is commonly used to prepare model membranes and to study lipid-protein interactions.

  • Monomethyl phosphatidylethanolamine lipid used in membrane and lipid research.
  • Purity approximately 98.0%.
  • Molecular formula C38H76NO8P; molecular weight 706.0 g/mol.
  • Supplied as a powder in small milligram pack sizes (1 mg, 5 mg, 10 mg).
  • Synonym: 1,2-NMeDPPE; CAS 3930-13-0.

Catalog No. 50-002-84672 Supplier Medchemexpress LLC Supplier No. HYW70310210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

16:0 Monomethyl PE (1,2-NMeDPPE) is a monomethylated phosphatidylethanolamine lipid supplied as a purified powder for laboratory research applications. It is commonly used to prepare model membranes and to study lipid-protein interactions.

  • Monomethyl phosphatidylethanolamine lipid used in membrane and lipid research.
  • Purity approximately 98.0%.
  • Molecular formula C38H76NO8P; molecular weight 706.0 g/mol.
  • Supplied as a powder in small milligram pack sizes (1 mg, 5 mg, 10 mg).
  • Synonym: 1,2-NMeDPPE; CAS 3930-13-0.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.