Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 16:0 monomethyl PE | 3930-13-0 | MFCD00042875 | 98.0% | 706.0 g/mol | C38H76NO8P | 5 MG
SDP

Catalog No. 5000284794
Encompass_Preferred

16:0 Monomethyl PE (1,2-NMeDPPE) is a monomethyl phosphatidylethanolamine lipid with two palmitoyl (16:0) acyl chains. Supplied as a purified research reagent, it is used in membrane biophysics, lipid signaling studies, and model membrane construction. Typically provided as a powder for laboratory use.

  • Monomethyl phosphatidylethanolamine with palmitoyl (16:0) chains.
  • Molecular formula C38H76NO8P.
  • Molecular weight 706.0 g/mol.
  • Purity 98.0%.
  • Supplied as a powder; handle per SDS storage and handling instructions.
  • Suitable for membrane model systems and lipidomics applications.

Catalog No. 50-002-84794 Supplier Medchemexpress LLC Supplier No. HYW7031025MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

16:0 Monomethyl PE (1,2-NMeDPPE) is a monomethyl phosphatidylethanolamine lipid with two palmitoyl (16:0) acyl chains. Supplied as a purified research reagent, it is used in membrane biophysics, lipid signaling studies, and model membrane construction. Typically provided as a powder for laboratory use.

  • Monomethyl phosphatidylethanolamine with palmitoyl (16:0) chains.
  • Molecular formula C38H76NO8P.
  • Molecular weight 706.0 g/mol.
  • Purity 98.0%.
  • Supplied as a powder; handle per SDS storage and handling instructions.
  • Suitable for membrane model systems and lipidomics applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.