Learn More
Avanti Polar Lipids 16:0 PC POWDER 500MG
Supplier:  Avanti Polar Lipids 850355P500MG
Encompass
16:0 PC, 1,2-Dipalmitoyl-sn-Glycero-3-Phosphocholine (DPPC), White Powder, Molecular Weight: 734.05, Molecular Formula: C40H80NO8P
Catalog No. NC9005555