Learn More
MSC Baldwin Outer Air Filter, 16 1/2 in Length, 1 ea
Catalog No. 5000505400
Encompass
The Baldwin PA2300 is an outer air filter element designed to protect engines from airborne contaminants. This axial seal filter ensures proper equipment function by preventing particulate matter from reaching the combustion chamber.
  • Axial seal design
  • Securely affixed metal end caps with a raised gasket to ensure a proper seal
  • Prevents leaks and ingress of contaminants
  • Cellulose/paper media material
  • Features 10 3/8 in outside diameter
  • Measures 16 1/2 in length
  • Includes attached gaskets
Catalog No. 50-005-05400 Supplier MSC Supplier No. 10940005
The Baldwin PA2300 is an outer air filter element designed to protect engines from airborne contaminants. This axial seal filter ensures proper equipment function by preventing particulate matter from reaching the combustion chamber.
  • Axial seal design
  • Securely affixed metal end caps with a raised gasket to ensure a proper seal
  • Prevents leaks and ingress of contaminants
  • Cellulose/paper media material
  • Features 10 3/8 in outside diameter
  • Measures 16 1/2 in length
  • Includes attached gaskets