Learn More
Avanti Polar Lipids 16:1 (Δ9-Cis) PC, 4724-96-3, MFCD00214374, chloroform, 25mg, 10mg/mL 2.5mL
Catalog No. NC1650138
Encompass
Molecular Formula C40H76NO8P, Purity >99%, Formula Weight 730.007, Storage Temperature -20°C, 1,2-dipalmitoleoyl-sn-glycero-3-phosphocholine
Catalog No. NC1650138 Supplier Avanti Polar Lipids Supplier No. 850358C25MG
Molecular Formula C40H76NO8P, Purity >99%, Formula Weight 730.007, Storage Temperature -20°C, 1,2-dipalmitoleoyl-sn-glycero-3-phosphocholine