Learn More
MSC Thomas & Betts RB14-6F #6 Stud, 18 to 14 AWG Compatible, Partially Insulated, Crimp Connection, Standard Fork Terminal 0.31" Fork Width, Nylon Insulation, Blue, 0.87" Overall Length, 600 Volts, Copper Contacts
Catalog No. 501296596
Encompass
Thomas & Betts RB14-6F #6 Stud, 18 to 14 AWG Compatible, Partially Insulated, Crimp Connection, Standard Fork Terminal 0.31" Fork Width, Nylon Insulation, Blue, 0.87" Overall Length, 600 Volts, Copper Contacts
Catalog No. 50-129-6596 Supplier MSC Supplier No. 54063532
Thomas & Betts RB14-6F #6 Stud, 18 to 14 AWG Compatible, Partially Insulated, Crimp Connection, Standard Fork Terminal 0.31" Fork Width, Nylon Insulation, Blue, 0.87" Overall Length, 600 Volts, Copper Contacts