Learn More
Cml Supply 16.15 BLUNT TIP 1000/BAG
Catalog No. NC9165286
Encompass_Preferred
Blunt Tipped Industrial Dispensing Needles, 16g x 1- 1/2 in. Luer Lock Poly Hub,1000/BG
Catalog No. NC9165286 Supplier Cml Supply Supplier No. 90116150M
Blunt Tipped Industrial Dispensing Needles, 16g x 1- 1/2 in. Luer Lock Poly Hub,1000/BG