Learn More
Medchemexpress LLC (±)16,17-dihydroxy-4Z,7Z,10Z,13Z,19Z-docosapentaenoic acid | 1345275-27-5 | 99.0% | 100 UG
SDP
Catalog No. 5000233259
Encompass_Preferred
(±)16(17)-DiHDPA is an epoxygenase metabolite of docosahexaenoic acid (DHA) that inhibits platelet aggregation at concentrations below those affecting thromboxane synthesis. It is supplied as a high-purity research compound for biochemical and pharmacological studies.
  • Epoxygenase metabolite of docosahexaenoic acid (DHA).
  • Inhibits platelet aggregation at low concentrations.
  • High purity (99.0%).
  • Molecular formula C22H34O4; molecular weight 362.50 g·mol⁻¹.
  • Supplied in small microgram quantities for research use.
Catalog No. 50-002-33259 Supplier Medchemexpress LLC Supplier No. HY120442100UG
(±)16(17)-DiHDPA is an epoxygenase metabolite of docosahexaenoic acid (DHA) that inhibits platelet aggregation at concentrations below those affecting thromboxane synthesis. It is supplied as a high-purity research compound for biochemical and pharmacological studies.
  • Epoxygenase metabolite of docosahexaenoic acid (DHA).
  • Inhibits platelet aggregation at low concentrations.
  • High purity (99.0%).
  • Molecular formula C22H34O4; molecular weight 362.50 g·mol⁻¹.
  • Supplied in small microgram quantities for research use.