Learn More
Medchemexpress LLC 16α,17α-Epoxyprogesterone | 1097-51-4 | 25 G
SDP
Catalog No. 5000422041
Encompass_Preferred
16α,17α-Epoxyprogesterone is an important steroid that serves as an intermediate for many hormonal drugs. It can also biotransform into 11α-hydroxy-16α,17α-epoxyprogesterone, another significant intermediate for manufacturing additional compounds. This product is for research use only.
  • Acts as an intermediate for numerous hormonal drugs
  • Can biotransform into other significant intermediates
  • Suitable for research use
Catalog No. 50-004-22041 Supplier Medchemexpress LLC Supplier No. 25GHYN3994
16α,17α-Epoxyprogesterone is an important steroid that serves as an intermediate for many hormonal drugs. It can also biotransform into 11α-hydroxy-16α,17α-epoxyprogesterone, another significant intermediate for manufacturing additional compounds. This product is for research use only.
  • Acts as an intermediate for numerous hormonal drugs
  • Can biotransform into other significant intermediates
  • Suitable for research use