Learn More
Cayman Chemical 16 17EpDPA Fatty Acids and Der
SDP
Catalog No. 502888515
Encompass_Preferred
A DHA metabolite derived via epoxidation of the 16,17-double bond; epoxygenase metabolites of DHA have been detected in a mouse model of inflammation
Catalog No. 50-288-8515 Supplier Cayman Chemical Supplier No. 1017450 UG
A DHA metabolite derived via epoxidation of the 16,17-double bond; epoxygenase metabolites of DHA have been detected in a mouse model of inflammation