Learn More
Cayman Chemical 16 17EpDPA Fatty Acids and Der
SDP
Supplier:  Cayman Chemical 10174100 UG
Encompass_Preferred
A DHA metabolite derived via epoxidation of the 16,17-double bond; epoxygenase metabolites of DHA have been detected in a mouse model of inflammation
Catalog No. 50-288-8830