Learn More
Cayman Chemical 16 17EpDPA Fatty Acids and Der
Supplier: Cayman Chemical 10174500 UG
A DHA metabolite derived via epoxidation of the 16,17-double bond; epoxygenase metabolites of DHA have been detected in a mouse model of inflammation