Learn More
Electron Microscopy Sciences 16/32 Slide Glass Rack With Handle
SDP
Catalog No. 5030251
Encompass_Preferred
This glass rack holds up to 26 single slides, or 32 slides back-to-back. The rack is made from soda-lime.
Catalog No. 50-302-51 Supplier Electron Microscopy Sciences Supplier No. 7031235
This glass rack holds up to 26 single slides, or 32 slides back-to-back. The rack is made from soda-lime.