Learn More
Chemglass Life Sciences Replacement Nuts for 16/35 PTFE Plug
SDP
Catalog No. 50872692
Encompass_Preferred
Replacement Nuts for 16/35 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.
Catalog No. 50-872-692 Supplier Chemglass Life Sciences Supplier No. CG45718
Replacement Nuts for 16/35 PTFE Plug. Replacement components for 1:5 taper PTFE plugs. O-rings are Viton, nuts are green, and washers are white.