Learn More
Starna Cells Inc 16.50-Q-10/50UL CUVETTE
SDP
Catalog No. NC9800666
Encompass_Preferred
Sub-Micro Cells Type 16, Small Sample Chamber, 2mm Wide, in Fully Masked Standard Exterior Size Cell, Window material: Spectrosil Quartz or equivalent, Range: 170 to 2700nm
Catalog No. NC9800666 Supplier Starna Cells Inc Supplier No. 16.50Q10/Z15
Sub-Micro Cells Type 16, Small Sample Chamber, 2mm Wide, in Fully Masked Standard Exterior Size Cell, Window material: Spectrosil Quartz or equivalent, Range: 170 to 2700nm