Learn More
Fine Science Tools Surgical Scissors, Sharp, Stainless Steel, 16.5 cm
SDP
Catalog No. NC9002311
Encompass
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 53mm/ Alloy / Material: Stainless Steel/ Length: 16.5cm/ Serrated: No
Catalog No. NC9002311 Supplier Fine Science Tools Supplier No. 1400216
Tip Shape: Straight/ Tips: Sharp-Sharp/ Cutting Edge: 53mm/ Alloy / Material: Stainless Steel/ Length: 16.5cm/ Serrated: No