Learn More
Sarstedt Inc 16.8x105mm Tube, 14mL, Round Bottom, Polypropylene, 1000/Case
Catalog No. NC9164146 Shop All Sarstedt Inc. Products
Encompass_Preferred
16.8x105mm Tube 14mL Round Bottom Polypropylene No Cap
Catalog No. NC9164146 Supplier Sarstedt Inc Supplier No. 55.538
16.8x105mm Tube 14mL Round Bottom Polypropylene No Cap