Learn More
Sarstedt Inc 16.8x95mm Tube, 13mL, Round Bottom, Polypropylene, 1000/Case
Catalog No. NC9021580 Shop All Sarstedt Inc. Products
Encompass_Preferred
16.8x95mm Tube 13mL Round Bottom Polypropylene No Cap
Catalog No. NC9021580 Supplier Sarstedt Inc Supplier No. 55.518
16.8x95mm Tube 13mL Round Bottom Polypropylene No Cap