Learn More
Medchemexpress LLC 16 alpha-hydroxydehydroisoandrosterone | 1232-73-1 | 98.4% | 304.42 | 1 MG
SDP
Catalog No. 5000439762
Encompass_Preferred
16a-Hydroxydehydroisoandrosterone (16α-Hydroxy-DHEA) is a metabolite of the endogenous steroid hormone dehydroepiandrosterone. It is formed from dehydroepiandrosterone via 16-hydroxylation by cytochrome P450 (CYP) isoforms CYP3A4 and CYP3A5 in adult human liver microsomes, and by fetal recombinant CYP3A7. It serves as a precursor to fetal estrogens, including estriol.
  • Metabolite of endogenous steroid hormone.
  • Formed by specific cytochrome P450 isoforms.
  • Precursor to fetal estrogens.
  • For research use only.
Catalog No. 50-004-39762 Supplier Medchemexpress LLC Supplier No. HY113031S1500UG
16a-Hydroxydehydroisoandrosterone (16α-Hydroxy-DHEA) is a metabolite of the endogenous steroid hormone dehydroepiandrosterone. It is formed from dehydroepiandrosterone via 16-hydroxylation by cytochrome P450 (CYP) isoforms CYP3A4 and CYP3A5 in adult human liver microsomes, and by fetal recombinant CYP3A7. It serves as a precursor to fetal estrogens, including estriol.
  • Metabolite of endogenous steroid hormone.
  • Formed by specific cytochrome P450 isoforms.
  • Precursor to fetal estrogens.
  • For research use only.