Learn More
GECCO BIOTECH B V 16 BVMOS SELECTED TO COVER
Catalog No. NC4026973
Encompass
16 BVMOs, selected to cover a wide substrate scope. Supplied as cell-free extract, lyophilized, 50 mg of each variant. Produced on demand, formatting (sample amount per plate & plate type) is flexible. Includes cofactor regeneration mix (cofactor, regenerating enzyme and NADP+).
Catalog No. NC4026973 Supplier GECCO BIOTECH B V Supplier No. BVMOKIT1
16 BVMOs, selected to cover a wide substrate scope. Supplied as cell-free extract, lyophilized, 50 mg of each variant. Produced on demand, formatting (sample amount per plate & plate type) is flexible. Includes cofactor regeneration mix (cofactor, regenerating enzyme and NADP+).