Learn More
Integra Biosciences Corp 16 Channel EVOLVE
SDP
Catalog No. NC1695939
Encompass
Traditional pipettes utilize a single rotating plunger to set the volumes. This approach makes frequent adjustment of volumes tedious, especially when switching from lower to higher volumes. To overcome this pain point, the EVOLVE manual pipette has been designed with three adjustable dials to rapidly set volumes. This revolutionary approach reduces repetitive stress injuries and allows users to set volumes in a fraction of the time it takes with a traditional manual pipette.
Catalog No. NC1695939 Supplier Integra Biosciences Corp Supplier No. 3045
Traditional pipettes utilize a single rotating plunger to set the volumes. This approach makes frequent adjustment of volumes tedious, especially when switching from lower to higher volumes. To overcome this pain point, the EVOLVE manual pipette has been designed with three adjustable dials to rapidly set volumes. This revolutionary approach reduces repetitive stress injuries and allows users to set volumes in a fraction of the time it takes with a traditional manual pipette.