Learn More
V&P Scientific Inc ASPIRATION MANIFOLD, 16 Channel, 384 Well, Standard, 20 Gauge, SS Tubes, 14mm Long, 4.5mm Spacing, Polycarbonate
Catalog No. NC1471073
Encompass
ASPIRATION MANIFOLD, 16 Channel, 384 Well, Standard Plate, 20 Gauge, Stainless Steel Tubes, 14mm Long, 4.5mm Center Spacing, Polycarbonate, Female Luer Lok
Catalog No. NC1471073 Supplier V&P Scientific Inc Supplier No. VP 189MV
ASPIRATION MANIFOLD, 16 Channel, 384 Well, Standard Plate, 20 Gauge, Stainless Steel Tubes, 14mm Long, 4.5mm Center Spacing, Polycarbonate, Female Luer Lok