Learn More
Cayman Chemical 16-doxyl StearIc AcId 10mg
SDP
Catalog No. 502981720
Encompass_Preferred
A spin trap for membrane fluidity studies
Catalog No. 50-298-1720 Supplier Cayman Chemical Supplier No. 2035510 MG
A spin trap for membrane fluidity studies