Learn More
Medchemexpress LLC 16-doxylstearic acid (free radical) | 53034-38-1 | MFCD00010066 | 98.0% | 384.57 g/mol | C22H42NO4 | 10 MG
SDP
Catalog No. 5000281630
Encompass_Preferred
16-Doxylstearic acid is a spin-labeled stearic acid used as a lipophilic probe to study membrane structure, dynamics, and protein-lipid interactions. Supplied as a stable nitroxide free radical, it is intended for use in electron paramagnetic resonance (EPR) and other spectroscopic techniques in biophysical and biochemical research.
  • Spin-labeled fatty acid probe for membrane studies.
  • Suitable for electron paramagnetic resonance (EPR) spectroscopy.
  • Hydrophobic nitroxide label reports local lipid environment.
  • Available in small research quantities for laboratory use.
  • High purity for reproducible biophysical measurements.
Catalog No. 50-002-81630 Supplier Medchemexpress LLC Supplier No. HYW25097110MG
Add to Cart
Add to Cart
16-Doxylstearic acid is a spin-labeled stearic acid used as a lipophilic probe to study membrane structure, dynamics, and protein-lipid interactions. Supplied as a stable nitroxide free radical, it is intended for use in electron paramagnetic resonance (EPR) and other spectroscopic techniques in biophysical and biochemical research.
  • Spin-labeled fatty acid probe for membrane studies.
  • Suitable for electron paramagnetic resonance (EPR) spectroscopy.
  • Hydrophobic nitroxide label reports local lipid environment.
  • Available in small research quantities for laboratory use.
  • High purity for reproducible biophysical measurements.