Learn More
Cayman Chemical 16-epI LatrunculIn B 100ug
SDP
Catalog No. 502982974
Encompass_Preferred
A naturally derived macrolide that inhibits actin polymerization in vitro at 5-10 UG/ml; demonstrates cytotoxic activity in mouse KA31T and NIH3T3 tumor cells (GI50s = 1 and 4 UG/ml, respectively) and antiviral activity against HSV-1 (ED50 = 1 UG/ml)
Catalog No. 50-298-2974 Supplier Cayman Chemical Supplier No. 19608100 UG
A naturally derived macrolide that inhibits actin polymerization in vitro at 5-10 UG/ml; demonstrates cytotoxic activity in mouse KA31T and NIH3T3 tumor cells (GI50s = 1 and 4 UG/ml, respectively) and antiviral activity against HSV-1 (ED50 = 1 UG/ml)