Learn More
Cayman Chemical 16-Epiestriol, Purity: ≥90%, CAS: 547-81-9, 100 mg
SDP
Catalog No. 502949516
Encompass_Preferred
A metabolite of estrone; inhibits the growth of carbapenem-resistant A. baumannii at 200 µg/ml; inhibits carrageenan-induced paw edema in rats at 20 mg/kg; does not increase plasma or liver glucose levels in adrenalectomized rats at 240 µg/animal
Catalog No. 50-294-9516 Supplier Cayman Chemical Supplier No. 33455100 MG
A metabolite of estrone; inhibits the growth of carbapenem-resistant A. baumannii at 200 µg/ml; inhibits carrageenan-induced paw edema in rats at 20 mg/kg; does not increase plasma or liver glucose levels in adrenalectomized rats at 240 µg/animal