Learn More
Cayman Chemical 16-EpIestrIol 50mg
SDP
Catalog No. 502948516
Encompass_Preferred
A metabolite of estrone; inhibits the growth of carbapenem-resistant A. baumannii at 200 UG/ml; inhibits carrageenan-induced paw edema in rats at 20 mg/kg; does not increase plasma or liver glucose levels in adrenalectomized rats at 240 UG/animal
Catalog No. 50-294-8516 Supplier Cayman Chemical Supplier No. 3345550 MG
A metabolite of estrone; inhibits the growth of carbapenem-resistant A. baumannii at 200 UG/ml; inhibits carrageenan-induced paw edema in rats at 20 mg/kg; does not increase plasma or liver glucose levels in adrenalectomized rats at 240 UG/animal