Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Ted Pella Inc Paraformaldehyde, 16% | 50-00-0 | MFCD00003274 |100ml
SDP

Catalog No. NC0528893
Encompass

Paraformaldehyde, 16% | MW: 30.026 | 50-00-0 | MFCD00003274 |100ml

Catalog No. NC0528893 Supplier Ted Pella Inc Supplier No. 18505100
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Paraformaldehyde, 16% | MW: 30.026 | 50-00-0 | MFCD00003274 |100ml

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.