Learn More
Cell Signaling Technology 16% Formaldehyde, Methanol-Free, 50 mL
SDP
Catalog No. 502050425
Encompass
This methanol-free 16% (w/v) formaldehyde solution is a high-quality fixative agent optimized for use in fluorescent immunocytochemical and flow cytometry assays. It is prepared from premium paraformaldehyde and packaged under an inert nitrogen atmosphere to ensure stability. The product is supplied in amber glass vials featuring dual access points, allowing for both bulk access via screw cap and precise small-volume extraction via needle and syringe through a silicone top to maximize shelf life.
  • Methanol-free formulation
  • Prepared from high-quality paraformaldehyde
  • Stable for twelve months when stored at room temperature
  • Packaged under inert nitrogen atmosphere
  • Amber glass vial with two access points for flexible delivery
  • Compatible with standard immunofluorescence and flow cytometry workflows
Catalog No. 50-205-0425 Supplier Cell Signaling Technology Supplier No. 12606S
This methanol-free 16% (w/v) formaldehyde solution is a high-quality fixative agent optimized for use in fluorescent immunocytochemical and flow cytometry assays. It is prepared from premium paraformaldehyde and packaged under an inert nitrogen atmosphere to ensure stability. The product is supplied in amber glass vials featuring dual access points, allowing for both bulk access via screw cap and precise small-volume extraction via needle and syringe through a silicone top to maximize shelf life.
  • Methanol-free formulation
  • Prepared from high-quality paraformaldehyde
  • Stable for twelve months when stored at room temperature
  • Packaged under inert nitrogen atmosphere
  • Amber glass vial with two access points for flexible delivery
  • Compatible with standard immunofluorescence and flow cytometry workflows