Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Vikan™ 16 ft. Fiberglass Extension Handle with Bottle

Catalog No. 17227582 Shop All Remco Products
Change view
Click to view available options
Color:
White
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
17-227-582 White
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 17-227-582 Supplier Vikan™ Supplier No. 6269B

16 ft. Fiberglass Extension Handle with Bottle is suitable for reaching tall ductwork to remove condensation before it falls onto food or onto expensive machinery.

  • This fiberglass extension pole extends to 16 feet
  • Drains into the included bottle part number RP6946
  • The pole is designed to work in conjunction with our condensation collection squeegees to eliminate dripping water that could contaminate food product or cause corrosion on costly equipment
  • The ceiling squeegee system provides a more sophisticated and effective option than rags and sponges
  • Color-coded

Specifications

Product Type Handle
Color-coded Yes
Sterility Non-sterile
Color White
Length (English) 102.63 to 192.5 in.
Material Fiberglass
Quantity 1 Ea.
Width (English) 1.2 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.