Learn More
MSC PRO-SAFE PSG1540 16 Gallon, 0.4 GPM Flow Rate at 30 PSI, Gravity Fed Polyethylene, Portable Eye Wash Station 15 Min Duration, 22 Inch Wide x 20 Inch Deep x 20-1/2 Inch High
Catalog No. NC1545630
Encompass
PRO-SAFE PSG1540 16 Gallon, 0.4 GPM Flow Rate at 30 PSI, Gravity Fed Polyethylene, Portable Eye Wash Station 15 Min Duration, 22 Inch Wide x 20 Inch Deep x 20-1/2 Inch High
This item is not returnable. View return policy
Catalog No. NC1545630 Supplier MSC Supplier No. 86205341
PRO-SAFE PSG1540 16 Gallon, 0.4 GPM Flow Rate at 30 PSI, Gravity Fed Polyethylene, Portable Eye Wash Station 15 Min Duration, 22 Inch Wide x 20 Inch Deep x 20-1/2 Inch High