Learn More
Cayman Chemical 16-hydroxy HexadecnoIc AcId 5g
SDP
Catalog No. 502895373
Encompass_Preferred
A metabolite of the saturated fatty acid palmitic acid (16:0) that has been hydroxylated on its terminal (ω) carbon; produced by ω-hydroxylation of palmitic acid by cytochrome P450 in both plants and animals; a common component of cutin in plants
Catalog No. 50-289-5373 Supplier Cayman Chemical Supplier No. 90007895 G
A metabolite of the saturated fatty acid palmitic acid (16:0) that has been hydroxylated on its terminal (ω) carbon; produced by ω-hydroxylation of palmitic acid by cytochrome P450 in both plants and animals; a common component of cutin in plants