Learn More
Medchemexpress LLC 16α-Hydroxydehydroepiandrosterone-d6 | 97453-29-7 | 80.0% | 310.46 | 1 MG
SDP
Catalog No. 5000439856
Encompass_Preferred
16α-Hydroxydehydroepiandrosterone-d6 (16α-Hydroxy-DHEA-d6) is a deuterium-labeled 16α-Hydroxydehydroepiandrosterone, intended for research use only. This compound is valuable as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is increasingly recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-39856 Supplier Medchemexpress LLC Supplier No. HY113031S1MG
16α-Hydroxydehydroepiandrosterone-d6 (16α-Hydroxy-DHEA-d6) is a deuterium-labeled 16α-Hydroxydehydroepiandrosterone, intended for research use only. This compound is valuable as a tracer and an internal standard for quantitative analysis using techniques such as NMR, GC-MS, or LC-MS. Deuteration, the incorporation of stable heavy isotopes of hydrogen, carbon, and other elements into drug molecules, is increasingly recognized for its potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS