Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 16α-Hydroxyestrone | 566-76-7 | 99.7% | 286.37 | 1 MG
SDP

Catalog No. 5000438404
Encompass_Preferred

16α-Hydroxyestrone (16αOHE) is a major metabolite of Estradiol. It can be used for the research of metabolic diseases. Increasing levels of active estrogen metabolites, such as 16α-Hydroxyestrone and E3, are associated with increased bone mineral density.

  • Endogenous metabolite
  • Steroid structure
  • Appearance: solid, white to off-white
  • Solubility: ≥ 250 mg/mL in DMSO

Catalog No. 50-004-38404 Supplier Medchemexpress LLC Supplier No. HY1133441MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

16α-Hydroxyestrone (16αOHE) is a major metabolite of Estradiol. It can be used for the research of metabolic diseases. Increasing levels of active estrogen metabolites, such as 16α-Hydroxyestrone and E3, are associated with increased bone mineral density.

  • Endogenous metabolite
  • Steroid structure
  • Appearance: solid, white to off-white
  • Solubility: ≥ 250 mg/mL in DMSO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.